Anti-MOG Mouse Antibody

Leverandør: Apollo Scientific
BITP2068-1MG
APOSBITP2068-1MGEA 0 NOK
APOSBITP2068-1MG
Anti-MOG Mouse Antibody
Antistoffer
Bestill nå

SPESIFIKASJONER

Antigensymbol MOG
Antigen-navn Myelin Oligodendrocyte Glycoprotein
Environmentally Preferable
Reaktivitet
Vert Mouse
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR