Antimon(III)klorid 99,9%

Leverandør: Apollo Scientific
Danger

Synonymer: Antimony trichloride

IN1108-100G IN1108-25G
APOSIN1108-100GEA 3750 NOK
APOSIN1108-100G APOSIN1108-25G
Antimon(III)klorid 99,9%
Antimon(III)klorid
Formel: SbCl₃
MW: 228.12 g/mol
Kokepunkt: 283 °C
Smeltepunkt: 73.4 °C
Tetthet: 3.14 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
EINECS: 233-047-2
FN: 1733
ADR: 8 II
Merck Indeks: 13,00713

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR