Antimony(III) isopropoxide 99,9%

Leverandør: Apollo Scientific

IN1120-10G
APOSIN1120-10GEA 3500 NOK
APOSIN1120-10G
Antimony(III) isopropoxide 99,9%
Antimony(III) isopropoxide
Formel: C₉H₂₁O₃Sb
Kokepunkt: 68 °C/3mmHg
Tetthet: 1,307 g/cm³ (25 °C)
Flash punkt: 170 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00143623
CAS-nummer: 18770-47-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR