Kobber (II) acetat monohydrat 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

IN1533-1KG
APOSIN1533-1KGEA 1975 NOK
APOSIN1533-1KG
Kobber (II) acetat monohydrat 98%
Kobber (II) acetat monohydrat

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

Formel: (H₃CCOO)₂Cu·H₂O
Smeltepunkt: 115 °C
Tetthet: 1,882 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149570
CAS-nummer: 6046-93-1
EINECS: 205-553-3
FN: 3077
ADR: 9+EHS,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR