Nickel (II) klorid heksahydrat 99,95%

Leverandør: Apollo Scientific
Danger

IN2683-1KG IN2683-250G
APOSIN2683-1KGEA 4200 NOK
APOSIN2683-1KG APOSIN2683-250G
Nickel (II) klorid heksahydrat 99,95%
Nickel (II) klorid heksahydrat
Formel: NiCl₂·6H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149809
CAS-nummer: 7791-20-0
FN: 3288
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR