Nikkel(II)nitrat heksahydrat 99.995%

Leverandør: Apollo Scientific
Danger

IN2698-250G IN2698-50G
APOSIN2698-250GEA 3575 NOK
APOSIN2698-250G APOSIN2698-50G
Nikkel(II)nitrat heksahydrat 99.995%
Nikkel(II)nitrat heksahydrat
Formel: Ni(NO₃)₂·6H₂O
Kokepunkt: 137 °C
Tetthet: 2,05 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149805
CAS-nummer: 13478-00-7
EINECS: 236-068-5
FN: 2725
ADR: 5.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR