Kaliumsorbat 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

IN2901-1KG
APOSIN2901-1KGEA 930 NOK
APOSIN2901-1KG
Kaliumsorbat 98%
Kaliumsorbat
Formel: CH₃CH=CHCH=CHCOOK
MW: 150,22 g/mol
Smeltepunkt: 210 °C (dec.)
Tetthet: 1,36 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016546
CAS-nummer: 24634-61-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR