Kaliumjodid

Leverandør: Apollo Scientific
Warning

IN2918-1KG IN2918-250G
APOSIN2918-1KGEA 2250 NOK
APOSIN2918-1KG APOSIN2918-250G
Kaliumjodid
Kaliumjodid
Formel: Kl
Kokepunkt: 1330 °C
Smeltepunkt: 681 °C
Tetthet: 3,13 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011405
CAS-nummer: 7681-11-0
EINECS: 231-659-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR