Ytterbium(III) chloride hydrate 99.9+%

Leverandør: Apollo Scientific
Warning

IN3799-10G
APOSIN3799-10GEA 715 NOK
APOSIN3799-10G
Ytterbium(III) chloride hydrate 99.9+%
Ytterbium(III) chloride hydrate
Formel: Cl₃Yb·H₂O
Smeltepunkt: 150…155 °C
Tetthet: 2,575 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149884
CAS-nummer: 19423-87-1
EINECS: 233-800-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR