(S)-(+)-Camptothecin

Leverandør: Apollo Scientific
Danger

Synonymer: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

OR014824-1G OR014824-5G
APOSOR014824-1GEA 340 NOK
APOSOR014824-1G APOSOR014824-5G
(S)-(+)-Camptothecin
(S)-(+)-Camptothecin
Formel: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
Smeltepunkt: 260 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00081076
CAS-nummer: 7689-03-4
FN: 1544
ADR: 6.1 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR