Imidazo[1,2-a]pyrimidine

Leverandør: Apollo Scientific
Warning

OR01549-5G
APOSOR01549-5GEA 840 NOK
APOSOR01549-5G
Imidazo[1,2-a]pyrimidine
Imidazo[1,2-a]pyrimidine
Formel: C₆H₅N₃
MW: 119,1200 g/mol
Smeltepunkt: 127…129 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06245377
CAS-nummer: 274-95-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR