4'-Bromoacetophenone

Leverandør: Apollo Scientific
Warning

Synonymer: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

OR01684-100G
APOSOR01684-100GEA 270 NOK
APOSOR01684-100G
4'-Bromoacetophenone
4'-Bromoacetophenone
Formel: C₈H₇BrO
MW: 199,05 g/mol
Kokepunkt: 255…256 °C
Smeltepunkt: 49…53 °C
Tetthet: 1,647 g/cm³ (25 °C)
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000105
CAS-nummer: 99-90-1
EINECS: 202-799-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR