Ethyl-3,5-dichloro-4-hydroxybenzoate

Leverandør: Apollo Scientific
Warning

Synonymer: Ethyl 3,5-dichloro-4-hydroxybenzoate

OR0451-1G
APOSOR0451-1GEA 270 NOK
APOSOR0451-1G
Ethyl-3,5-dichloro-4-hydroxybenzoate
Ethyl-3,5-dichloro-4-hydroxybenzoate
Formel: C₉H₈Cl₂O₃
MW: 235,07 g/mol
Smeltepunkt: 110…112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016420
CAS-nummer: 17302-82-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR