Tris(dimethylamino)phosphine

Leverandør: Apollo Scientific
Danger

OR0499-25G
APOSOR0499-25GEA 1400 NOK
APOSOR0499-25G
Tris(dimethylamino)phosphine
Tris(dimethylamino)phosphine
Formel: C₆H₁₈N₃P
Kokepunkt: 162…164 °C
Smeltepunkt: –44 °C
Tetthet: 0,898 g/cm³ (25 °C)
Flash punkt: 37 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008301
CAS-nummer: 1608-26-0
FN: 1993
ADR: 3,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR