Methyl adamant-1-ylacetate

Leverandør: Apollo Scientific
Warning

OR0596-10G OR0596-1G OR0596-5G
APOSOR0596-10GEA 4275 NOK
APOSOR0596-10G APOSOR0596-1G APOSOR0596-5G
Methyl adamant-1-ylacetate
Methyl adamant-1-ylacetate
Formel: C₁₃H₂₀O₂
MW: 208.297 g/mol
Kokepunkt: 86 °C (0.3 mm Hg)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01073581
CAS-nummer: 27174-71-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR