2,5-Dimetoksyanilin

Leverandør: Apollo Scientific
Danger

OR0958-100G
APOSOR0958-100GEA 270 NOK
APOSOR0958-100G
2,5-Dimetoksyanilin
2,5-Dimetoksyanilin
Formel: C₈H₁₁NO₂
MW: 153,18 g/mol
Kokepunkt: 140 °C/7mmHg
Smeltepunkt: 78…82 °C
Flash punkt: 130 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008368
CAS-nummer: 102-56-7
EINECS: 203-040-9
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR