(Dimethylamino)acetaldehyde dimethylacetal 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 2,2-Dimethoxy-N,N-dimethylethanamine, N-(2,2-Dimethoxyethyl)dimethylamine

OR10113-5ML
APOSOR10113-5MLEA 270 NOK
APOSOR10113-5ML
(Dimethylamino)acetaldehyde dimethylacetal 98%
(Dimethylamino)acetaldehyde dimethylacetal
Formel: C₆H₁₅NO₂
MW: 133.1888 g/mol
Kokepunkt: 64…66 °C/12mmHg
Tetthet: 0.9 g/cm³ (25 °C)
Flash punkt: 32 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010205
CAS-nummer: 38711-20-5
FN: 1993
ADR: 3 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR