2-(Hydroxymethyl)phenylboronic acid cyclic monoester 99%

Leverandør: Apollo Scientific
Warning

OR10502-1G
APOSOR10502-1GEA 270 NOK
APOSOR10502-1G
2-(Hydroxymethyl)phenylboronic acid cyclic monoester 99%
2-(Hydroxymethyl)phenylboronic acid cyclic monoester
Formel: C₇H₇BO₂
Smeltepunkt: 98…100 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01075677
CAS-nummer: 5735-41-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR