2-Bromo-p-toluidine

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene

OR1073-100G OR1073-25G
APOSOR1073-100GEA 540 NOK
APOSOR1073-100G APOSOR1073-25G
2-Bromo-p-toluidine
2-Bromo-p-toluidine
Formel: C₇H₈BrN
Kokepunkt: 120…122 °C/30mmHg
Smeltepunkt: 14…16 °C
Tetthet: 1,5 g/cm³ (25 °C)
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007635
CAS-nummer: 583-68-6
EINECS: 209-515-7
FN: 2810
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR