Ethyl imidazo[1,2-a]pyrimidine-3-carboxylate

Leverandør: Apollo Scientific

OR110094-1G
APOSOR110094-1GEA 1350 NOK
APOSOR110094-1G
Ethyl imidazo[1,2-a]pyrimidine-3-carboxylate
Ethyl imidazo[1,2-a]pyrimidine-3-carboxylate
Formel: C₉H₉N₃O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD10698087
CAS-nummer: 64951-07-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR