3-Iodoimidazo[1,2-a]pyridine-6-carbonitrile

Leverandør: Apollo Scientific
Warning

OR110208-1G
APOSOR110208-1GEA 1075 NOK
APOSOR110208-1G
3-Iodoimidazo[1,2-a]pyridine-6-carbonitrile
3-Iodoimidazo[1,2-a]pyridine-6-carbonitrile
Formel: C₈H₄IN₃
MW: 269,042 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06739236
CAS-nummer: 885276-13-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR