2-(3-Nitrophenyl)-1,3,4-oxadiazole

Leverandør: Apollo Scientific

OR110325-1G
APOSOR110325-1GEA 0 NOK
APOSOR110325-1G
2-(3-Nitrophenyl)-1,3,4-oxadiazole
2-(3-Nitrophenyl)-1,3,4-oxadiazole
Formel: C₈H₅N₃O₃
MW: 191,144 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085145
CAS-nummer: 5565-72-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR