3-[1,3,4]Oxadiazol-2-ylphenylamine

Leverandør: Apollo Scientific
Warning

OR110326-1G
APOSOR110326-1GEA 2700 NOK
APOSOR110326-1G
3-[1,3,4]Oxadiazol-2-ylphenylamine
3-[1,3,4]Oxadiazol-2-ylphenylamine
Formel: C₈H₇N₃O
MW: 161,16 g/mol
Smeltepunkt: 45…48 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00099501
CAS-nummer: 5378-35-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR