4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide

Leverandør: Apollo Scientific

OR110549-1G
APOSOR110549-1GEA 4875 NOK
APOSOR110549-1G
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide
Formel: C₁₁H₁₁N₃O₂
MW: 217,224 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18384923
CAS-nummer: 1427460-93-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR