tert-Butyl 2-[(3-bromobenzoyl)amino]ethylcarbamate

Leverandør: Apollo Scientific

OR110646-1G
APOSOR110646-1GEA 3050 NOK
APOSOR110646-1G
tert-Butyl 2-[(3-bromobenzoyl)amino]ethylcarbamate
tert-Butyl 2-[(3-bromobenzoyl)amino]ethylcarbamate
Formel: C₁₄H₁₉BrN₂O₃
MW: 343,216 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18804161
CAS-nummer: 1427460-40-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR