5-Bromo-o-toluidine

Leverandør: Apollo Scientific
Warning

Synonymer: 5-Bromo-2-methylaniline, 5-Bromo-o-toluidine, 2-Amino-4-bromotoluene

OR11111-5G
APOSOR11111-5GEA 270 NOK
APOSOR11111-5G
5-Bromo-o-toluidine
5-Bromo-o-toluidine
Formel: C₇H₈BrN
MW: 186.05 g/mol
Kokepunkt: 139 °C/17mmHg
Smeltepunkt: 30…33 °C
Tetthet: 1.49 g/cm³ (25 °C)
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00800678
CAS-nummer: 39478-78-9
EINECS: 254-467-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR