3-(2-Hydroxy-5-methylphenyl)pyrazole

Leverandør: Apollo Scientific
Warning

Synonymer: 4-methyl-2-(1h-pyrazol-3-yl)phenol

OR111446-1G
APOSOR111446-1GEA 1575 NOK
APOSOR111446-1G
3-(2-Hydroxy-5-methylphenyl)pyrazole
3-(2-Hydroxy-5-methylphenyl)pyrazole
Formel: C₁₀H₁₀N₂O
MW: 174.20 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03424535
CAS-nummer: 57148-86-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR