2-Bromo-1-(2-chloropyridin-4-yl)ethanone hydrobromide

Leverandør: Apollo Scientific
Danger

OR111611-1G
APOSOR111611-1GEA 0 NOK
APOSOR111611-1G
2-Bromo-1-(2-chloropyridin-4-yl)ethanone hydrobromide
2-Bromo-1-(2-chloropyridin-4-yl)ethanone hydrobromide
Formel: C₇H₅BrClNO·BrH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD13192035
CAS-nummer: 569667-89-6
FN: 2923
ADR: 8+6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR