2-Ethylphenylboronic acid

Leverandør: Apollo Scientific
Warning

Synonymer: (2-Ethylphenyl)metaboric acid, 2-Ethylbenzeneboronic acid, Dihydroxy(2-ethylphenyl)borane

OR11254-5G
APOSOR11254-5GEA 270 NOK
APOSOR11254-5G
2-Ethylphenylboronic acid
2-Ethylphenylboronic acid
Formel: C₈H₁₁BO₂
MW: 149,98 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02093075
CAS-nummer: 90002-36-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR