2,6-Dimethoxyaniline hydrochloride

Leverandør: Apollo Scientific

OR11968-1G
APOSOR11968-1GEA 950 NOK
APOSOR11968-1G
2,6-Dimethoxyaniline hydrochloride
2,6-Dimethoxyaniline hydrochloride
Formel: C₈H₁₂ClNO₂
MW: 189,6394 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09878313
CAS-nummer: 375397-36-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR