4-Propoxy-3-methylphenylboronic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 3-Methyl-4-propoxybenzeneboronic acid, (3-Methyl-4-propoxyphenyl)boronic acid

OR1201-1G OR1201-5G
APOSOR1201-1GEA 860 NOK
APOSOR1201-1G APOSOR1201-5G
4-Propoxy-3-methylphenylboronic acid
4-Propoxy-3-methylphenylboronic acid
Formel: C₁₀H₁₅BO₃
MW: 194.0400 g/mol
Smeltepunkt: 154…156 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06659872
CAS-nummer: 279262-88-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR