3,5-Dimethyl-4-propoxyphenylboronic acid

Leverandør: Apollo Scientific

Synonymer: (3,5-Dimethyl-4-propoxyphenyl)boronic acid

OR1203-100G OR1203-25G OR1203-5G
APOSOR1203-100GEA 6625 NOK
APOSOR1203-100G APOSOR1203-25G APOSOR1203-5G
3,5-Dimethyl-4-propoxyphenylboronic acid
3,5-Dimethyl-4-propoxyphenylboronic acid
Formel: C₁₁H₁₇BO₃
MW: 208.0619 g/mol
Smeltepunkt: 170…173 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05865178
CAS-nummer: 357611-51-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR