4-Methoxy-o-toluidine 98%

Leverandør: Apollo Scientific
Warning

OR12498-100G OR12498-25G
APOSOR12498-100GEA 915 NOK
APOSOR12498-100G APOSOR12498-25G
4-Methoxy-o-toluidine 98%
4-Methoxy-o-toluidine
Formel: C₈H₁₁NO
Kokepunkt: 248…249 °C
Smeltepunkt: 13…14 °C
Tetthet: 1,065 g/cm³ (25 °C)
Flash punkt: 124 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007735
CAS-nummer: 102-50-1
EINECS: 203-036-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR