Methyl-3-methyl-2-furoate 98%

Leverandør: Apollo Scientific

Synonymer: Methyl 3-methylfuran-2-carboxylate

OR12570-5G
APOSOR12570-5GEA 950 NOK
APOSOR12570-5G
Methyl-3-methyl-2-furoate 98%
Methyl-3-methyl-2-furoate
Formel: C₇H₈O₃
MW: 140,14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014110
CAS-nummer: 6141-57-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR