3-(4-Pyridyl)benzaldehyde 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 3-(Pyridin-4-yl)benzaldehyde, 3-Pyrid-4-ylbenzaldehyde

OR12579-1G
APOSOR12579-1GEA 0 NOK
APOSOR12579-1G
3-(4-Pyridyl)benzaldehyde 97%
3-(4-Pyridyl)benzaldehyde
Formel: C₁₂H₉NO
MW: 183,2060 g/mol
Smeltepunkt: 47…49 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02684105
CAS-nummer: 208190-04-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR