4-(Pyrimidin-5-yl)benzaldehyde

Leverandør: Apollo Scientific
Warning

OR12646-5G
APOSOR12646-5GEA 2075 NOK
APOSOR12646-5G
4-(Pyrimidin-5-yl)benzaldehyde
4-(Pyrimidin-5-yl)benzaldehyde
Formel: C₁₁H₈N₂O
MW: 184,1940 g/mol
Smeltepunkt: 145…149 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02684108
CAS-nummer: 198084-12-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR