1-Acetyladamantane 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-Adamantyl Methyl Ketone, Methyl 1-Adamantyl Ketone

OR1265-100G
APOSOR1265-100GEA 1850 NOK
APOSOR1265-100G
1-Acetyladamantane 98%
1-Acetyladamantane
Formel: C₁₂H₁₈O
MW: 178.28 g/mol
Smeltepunkt: 53…55 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00074739
CAS-nummer: 1660-04-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR