2,2-Diphenylethylamine 96%

Leverandør: Apollo Scientific

Synonymer: 1-Amino-2,2-diphenylethane, 2,2-Diphenylethanamine

OR12742-25G
APOSOR12742-25GEA 1325 NOK
APOSOR12742-25G
2,2-Diphenylethylamine 96%
2,2-Diphenylethylamine
Formel: C₁₄H₁₅N
MW: 197.28 g/mol
Smeltepunkt: 48…49 °C
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008143
CAS-nummer: 3963-62-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR