4-(4-Chlorophenoxy)piperidine hydrochloride 98%

Leverandør: Apollo Scientific
Warning

OR12798-1G
APOSOR12798-1GEA 2350 NOK
APOSOR12798-1G
4-(4-Chlorophenoxy)piperidine hydrochloride 98%
4-(4-Chlorophenoxy)piperidine hydrochloride
Formel: C₁₁H₁₅Cl₂NO
Smeltepunkt: 194…196 °C
Lagringstemperatur: Romtemperatur
CAS-nummer: 63843-53-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR