2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride

Leverandør: Apollo Scientific
Warning

Synonymer: 2-[(Piperidin-4-yl)oxy]-1,4-diazine dihydrochloride, 4-[(Pyrazin-2-yl)oxy]piperidine dihydrochloride

OR12809-50MG
APOSOR12809-50MGEA 500 NOK
APOSOR12809-50MG
2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride
2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride
Formel: C₉H₁₃N₃O·2ClH
MW: 252,141 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03840122
CAS-nummer: 950649-21-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR