Glykolsyre 98%

Leverandør: Apollo Scientific
Danger

OR13191-100G
APOSOR13191-100GEA 930 NOK
APOSOR13191-100G
Glykolsyre 98%
Glykolsyre
Formel: C₂H₄O₃
Smeltepunkt: 74…80 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004312
CAS-nummer: 79-14-1
EINECS: 201-180-5
FN: 3261
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR