5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol

Leverandør: Apollo Scientific
Warning

Synonymer: (3-Mercapto-5H-[1,2,4]triazino[5,6-b]indol-5-yl)acetic acid, 2-(3-Mercapto-5H-[1,2,4]triazino[5,6-b]indol-5-yl)acetic acid

OR13468-250MG
APOSOR13468-250MGEA 1550 NOK
APOSOR13468-250MG
5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol
5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol
Formel: C₁₁H₈N₄O₂S
MW: 260.27 g/mol
MDL nummer: MFCD01820802
CAS-nummer: 309283-89-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR