3-(Chloromethyl)-5-methylpyridine hydrochloride

Leverandør: Apollo Scientific
Warning

OR13615-25G
APOSOR13615-25GEA 1275 NOK
APOSOR13615-25G
3-(Chloromethyl)-5-methylpyridine hydrochloride
3-(Chloromethyl)-5-methylpyridine hydrochloride
Formel: C7H9Cl2N
MW: 178.0600 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11656303
CAS-nummer: 1007089-84-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR