6-Chloro-o-toluidine 97%

Leverandør: Apollo Scientific

Synonymer: 2-Chloro-6-methylaniline, 2-Amino-3-chlorotoluene

OR13655-100G
APOSOR13655-100GEA 1075 NOK
APOSOR13655-100G
6-Chloro-o-toluidine 97%
6-Chloro-o-toluidine
Formel: C₇H₈ClN
Kokepunkt: 215 °C
Smeltepunkt: 44540 °C
Tetthet: 1,152 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007679
CAS-nummer: 87-63-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR