4-Pyridineboronic acid hydrochloride

Leverandør: Apollo Scientific
Warning

Synonymer: Pyridin-4-ylboronic acid hydrochloride

OR14161-1G
APOSOR14161-1GEA 270 NOK
APOSOR14161-1G
4-Pyridineboronic acid hydrochloride
4-Pyridineboronic acid hydrochloride
Formel: C₅H₇BClNO₂
MW: 159,38 g/mol
Smeltepunkt: 254 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06201888
CAS-nummer: 913835-65-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR