4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid > 95%

Leverandør: Apollo Scientific
Warning

OR14294-250MG
APOSOR14294-250MGEA 0 NOK
APOSOR14294-250MG
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid > 95%
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid
Formel: C₁₂H₁₃NO₃
MW: 219.24 g/mol
Smeltepunkt: 169…171 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06411619
CAS-nummer: 2971-18-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR