N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine

Leverandør: Apollo Scientific
Warning

Synonymer: N-Boc-3-(2-naphthyl)-L-alanine, (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, (S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, Boc-2-Nal-OH

OR14598-25G
APOSOR14598-25GEA 1375 NOK
APOSOR14598-25G
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine
Formel: C₁₈H₂₁NO₄
MW: 315.37 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00079671
CAS-nummer: 58438-04-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR