2-Nitro-L-phenylalanine

Leverandør: Apollo Scientific
Warning

Synonymer: (2S)-2-Amino-3-(2-nitrophenyl)propionic acid, (2S)-2-Amino-3-(2-nitrophenyl)propanoic acid, (S)-2-Amino-3-(2-nitrophenyl)propanoic acid, H-Phe(2-NO₂)-OH

OR14705-250MG
APOSOR14705-250MGEA 270 NOK
APOSOR14705-250MG
2-Nitro-L-phenylalanine
2-Nitro-L-phenylalanine
Formel: C₉H₁₀N₂O₄
MW: 210.187 g/mol
Smeltepunkt: 226…227 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270360
CAS-nummer: 19883-75-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR