3-Bromo-5-(chloromethyl)pyridine hydrochloride 95+%

Leverandør: Apollo Scientific
Danger

Synonymer: Pyridine, 3-bromo-5-(chloromethyl)-, hydrochloride

OR14923-1G
APOSOR14923-1GEA 930 NOK
APOSOR14923-1G
3-Bromo-5-(chloromethyl)pyridine hydrochloride 95+%
3-Bromo-5-(chloromethyl)pyridine hydrochloride
Formel: C₆H₆BrCl₂N
MW: 242.925 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09260933
CAS-nummer: 39741-46-3
FN: 1759
ADR: 8 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR