3-Chloro-6-(2-methoxyphenyl)pyridazine

Leverandør: Apollo Scientific
Warning

OR14962-250MG
APOSOR14962-250MGEA 2500 NOK
APOSOR14962-250MG
3-Chloro-6-(2-methoxyphenyl)pyridazine
3-Chloro-6-(2-methoxyphenyl)pyridazine
Formel: C₁₁H₉ClN₂O
MW: 220,655 g/mol
Smeltepunkt: 122 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06801341
CAS-nummer: 949468-94-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR